• USA Home
  • 735051 - 2,4,6-Triisopropylphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
735051 Sigma-Aldrich

2,4,6-Triisopropylphenylboronic acid

Synonym: 2,4,6-Triisopropylbenzeneboronic acid

  • CAS Number 154549-38-9

  • Empirical Formula (Hill Notation) C15H25BO2

  • Molecular Weight 248.17

  •  MDL number MFCD02683099

  •  PubChem Substance ID 329763790

  •  NACRES NA.22

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents,
Quality Level   100
mp   167-172 °C
storage temp.   2-8°C
SMILES string   CC(C)c1cc(C(C)C)c(B(O)O)c(c1)C(C)C
InChI   1S/C15H25BO2/c1-9(2)12-7-13(10(3)4)15(16(17)18)14(8-12)11(5)6/h7-11,17-18H,1-6H3
InChI key   FGFYEKLWZBTLEW-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?