EMAIL THIS PAGE TO A FRIEND
736686 Sigma-Aldrich

Allyl carbamate

95%

Purchase

Properties

Related Categories Amides, Building Blocks, C2 to C5, C2 to C7, Carbonyl Compounds,
InChI Key   OCAAZRFBJBEVPS-UHFFFAOYSA-N
assay   95%
refractive index   n20/D 1.454
density   1.077 g/mL at 25 °C
storage temp.   −20°C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?