• USA Home
  • 736732 - 4-Bromopyrrole-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
736732 Sigma-Aldrich

4-Bromopyrrole-2-carboxylic acid

95%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrroles
InChI Key   YUWKEVJEMKKVGQ-UHFFFAOYSA-N
assay   95%
mp   183-189 °C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?