• USA Home
  • 737089 - 2-(Methylamino)pyridine-3-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
737089 Sigma-Aldrich

2-(Methylamino)pyridine-3-carboxylic acid

95%

Synonym: 2-Methylaminonicotinic acid

Purchase

Properties

Related Categories Building Blocks, C7, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   SMYTUBMIBZOXTH-UHFFFAOYSA-N
assay   95%
mp   250-255 °C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?