• USA Home
  • 73876 - Boc-D-Abu-OH (dicyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
73876 Sigma-Aldrich

Boc-D-Abu-OH (dicyclohexylammonium) salt

≥98.0% (CHN)

Synonym: (R)-2-Boc-amino)butyric acid (dicyclohexylammonium) salt

  • CAS Number 27494-47-9

  • Empirical Formula (Hill Notation) C9H17NO4 · C12H23N

  • Molecular Weight 384.55

  •  Beilstein/REAXYS Number 6032357

  •  MDL number MFCD00235886

  •  eCl@ss 32160406

  •  PubChem Substance ID 57652494

Purchase

Properties

Related Categories Alanine Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   ≥98.0% (CHN)
application(s)   peptide synthesis: suitable
storage temp.   2-8°C
SMILES string   C1CCC(CC1)NC2CCCCC2.CC[C@@H](NC(=O)OC(C)(C)C)C(O)=O
InChI   1S/C12H23N.C9H17NO4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-5-6(7(11)12)10-8(13)14-9(2,3)4/h11-13H,1-10H2;6H,5H2,1-4H3,(H,10,13)(H,11,12)/t;6-/m.1/s1
InChI key   HOSMYZXDFVUSCV-FCXZQVPUSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?