• USA Home
  • 74140 - trans-4-Nitrocinnamic acid

EMAIL THIS PAGE TO A FRIEND
74140 Sigma-Aldrich

trans-4-Nitrocinnamic acid

≥97.0%

Synonym: trans-3-(4-Nitrophenyl)acrylic acid

  • CAS Number 882-06-4

  • Empirical Formula (Hill Notation) C9H7NO4

  • Molecular Weight 193.16

  •  Beilstein/REAXYS Number 2049190

  •  EC Number 212-924-3

  •  MDL number MFCD00007381

  •  PubChem Substance ID 24886648

Purchase

Properties

assay   ≥97.0%
mp   ~290 °C (dec.)
SMILES string   OC(=O)\C=C\c1ccc(cc1)[N+]([O-])=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?