EMAIL THIS PAGE TO A FRIEND
745707 Sigma-Aldrich

Reactive silver ink

Synonym: Ag ink, Diamminesilver(I) acetate

Purchase

Properties

Related Categories 3D Printing Materials for Research and Development, Conductive Inks for 3D Printing, Inks for Printing Applications, Materials Science, Organic and Printed Electronics,
InChI Key   CQLFBEKRDQMJLZ-UHFFFAOYSA-M
assay   12-14 wt. % Ag basis
form   liquid
viscosity   10-12 cP
density   1.0367 g/L at 25 °C
storage temp.   2-8°C

Description

Packaging

25 mL in glass bottle

Warning

This product is for R&D use only; not for drug; household; or other uses. Please consult the Materials Safety Data Sheet for information regarding hazards and safe handling procedures.

Application

Reactive silver ink based on silver acetate complex is a particle-free silver ink that thermally decomposes to silver upon mild heating. It can be inkjet printed; spincoated and patterned using roll-to-roll processes for printing and patterning applications.

General description

Silver is a good conductor and is resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles). Nanosilver is used as an antibacterial material in filters for air and water purification, in textiles, and for wound treatment. Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts.

The product can be stored at room temperature for several months. However if the user plans on not using the ink for long periods of time; refrigerated storage is recommended for best shelf life.

Legal Information

Product of Electroninks Incorporated.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN2734 - class 8 - PG 2 - EHS - Amines, liquid, corrosive, flammable, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
116.6 °F
Flash Point(C) 
47 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

3D Printing of Carbon Fiber-Reinforced Composites

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries, most typically using a layer-by-layer fabrication process. 3D pri...
Zhenyu Bo* (Ph.D Candidate at Northwestern University) and Jia Choi*, PhD, Product Manager


*Materials Science Product Management Team, MilliporeSigma, Milwaukee, WI.
Keywords: Deposition, Nanomaterials, Nanotubes

Graphene Inks for Printed Electronics

The emerging field of printed electronics requires a suite of functional materials for applications including flexible and large-area displays,1 radio frequency identification tags,2 portable energy ...
Ethan B. Secor and Professor Mark C. Hersam

Department of Materials Science and Engineering and Department of Chemistry, Northwestern University, Evanston, IL 60208-3108 USA
Keywords: Chemical vapor deposition, Degradations, Deposition, Electronics, Environmental, Evaporation, Microscopy, Nanomaterials, Nanotubes, Nucleic acid annealing, Oxidations, Photovoltaics, Printed electronics, Semiconductor, Solvents

Nanoparticle-based Zinc Oxide Electron Transport Layers for Printed Organic Photodetectors

Gerardo Hernandez-Sosa,1,2* Ralph Eckstein,1,2 Tobias Rödlmeier,1,2 Uli Lemmer1,2,3 1Lichttechnisches Institut, Karlsruher Institut für Technologie, Engesserstrasse 13, 76131 Karlsruhe, Germany 2Inno...
Gerardo Hernandez-Sosa*, Ralph Eckstein, Tobias Rödlmeier, Uli Lemmer
Material Matters, 2016, 11.2
Keywords: Deposition, Electronics, Positron Emission Tomography, Recombination, Reductions, Solar cells, Thin film deposition

Reactive Silver Inks for High-Performance Printed Electronics

S. Brett Walker,1,2 Bok Yeop Ahn,3* Jennifer A. Lewis1,3,4 1Materials Science and Engineering Department University of Illinois at Urbana-Champaign, Urbana, IL 61801 2Electroninks Incorporated, Champ...
Keywords: Absorption, Adhesion, Decarboxylations, Deposition, Electronics, Ligands, Nucleic acid annealing, Oxidations, Positron Emission Tomography, Printed electronics, Ultraviolet-Visible spectroscopy

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?