• USA Home
  • 749338 - 5-Methylthiophene-2-sulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
749338 Sigma-Aldrich

5-Methylthiophene-2-sulfonyl chloride

97%

Purchase

Properties

Related Categories Building Blocks, C4 to C6, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   QGGRRHYGHGJEKP-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.569
density   1.432 g/mL at 25 °C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGIII
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?