• USA Home
  • 749826 - TMEDA (1,5-cyclooctadiene) rhodium(I)/dichloro(1,5-cyclooctadiene)rhodium(I) complex

EMAIL THIS PAGE TO A FRIEND
749826 Sigma-Aldrich

TMEDA (1,5-cyclooctadiene) rhodium(I)/dichloro(1,5-cyclooctadiene)rhodium(I) complex

Synonym: [Rh(TMEDA) (COD)]+[RhCl2(COD)]-

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Rhodium Catalysts More...
InChI Key   OLXDNJYVTHYWRC-YHSVTWOFSA-L
mp   220-227 °C

Description

Packaging

1 g in glass bottle

250 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?