• USA Home
  • 751995 - Potassium 5-formylthiophene-3-trifluoroborate

EMAIL THIS PAGE TO A FRIEND
751995 Sigma-Aldrich

Potassium 5-formylthiophene-3-trifluoroborate

95%

Synonym: Potassium 5-formylthiophen-3-yltrifluoroborate, Potassium trifluoro(5-formylthien-3-yl)borate

  • CAS Number 907604-61-9

  • Empirical Formula (Hill Notation) C5H3BF3KOS

  • Molecular Weight 218.05

  •  MDL number MFCD11052810

  •  PubChem Substance ID 329766257

  •  NACRES NA.22

Purchase

Properties

Related Categories Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Trifluoroborate Salts, Organometallic Reagents, Trifluoroborate Salts More...
assay   95%
mp   262-273 °C
SMILES string   [K+].F[B-](F)(F)c1csc(C=O)c1
InChI   1S/C5H3BF3OS.K/c7-6(8,9)4-1-5(2-10)11-3-4;/h1-3H;/q-1;+1
InChI key   COQNXWNZUJWEAO-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?