• USA Home
  • 752355 - 5-Bromo-3-methylindole

EMAIL THIS PAGE TO A FRIEND
752355 Sigma-Aldrich

5-Bromo-3-methylindole

97%

Synonym: 5-Bromo-3-methyl-1H-indole

  • CAS Number 10075-48-6

  • Empirical Formula (Hill Notation) C9H8BrN

  • Molecular Weight 210.07

  •  MDL number MFCD04038746

  •  PubChem Substance ID 329766281

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C7 to C9, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
Quality Level   100
assay   97%
mp   78-82 °C
storage temp.   2-8°C
SMILES string   Cc1c[nH]c2ccc(Br)cc12
InChI   1S/C9H8BrN/c1-6-5-11-9-3-2-7(10)4-8(6)9/h2-5,11H,1H3
InChI key   GDQXDVJFMLNXHX-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?