• USA Home
  • 752509 - Bismuth antimony telluride

EMAIL THIS PAGE TO A FRIEND
752509 Aldrich

Bismuth antimony telluride Green Alternative

beads, 99.99% trace metals basis

Synonym: Antimony bismuth telluride, BiSbTe

Purchase

Properties

Related Categories Alternative Energy, Chalcogenides, Chemical Synthesis, Enabling Products, Greener Alternative Products,
InChI Key   BOOKHSKFBCVCND-UHFFFAOYSA-N
assay   99.99% trace metals basis
form   beads
composition   Bi0.5Sb1.5Te3
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Description

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Thermoelectric properties of bismuth antimony telluride (BiSbTe) has been studied in detail.1 A study reports the preparation of p-type BiSbTe by melting solidification method in static magnetic field. Concentration of holes in p-type BiSbTe can be regulated by tellurium evaporation annealing technique. Thermoelectrical properties of dense p-type BiSbTe was compared to nanosized antimony-doped p-type BiSbTe. The properties were found to be enhanced.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Silicides: Promising Thermoelectric Materials - Material Matters Volume 6 Number 4

In recent years, the price of tellurium, a key component in the best-performing thermoelectric materials, has increased significantly, leading to the question, "Is it economically viable to produce t...
Mikhail I. Fedorov, Vladimir K. Zaitsev
Material Matters Volume 6 Number 4
Keywords: Applications, Gene expression, Melting, Methods, Nucleic acid annealing, Reductions, Semiconductor, Type

Silicides: Thermoelectric Materials

Mikhail I. Fedorov,* Vladimir K. Zaitsev Ioffe Physical-technical Institute, St. Petersburg, Russia *Email: m.fedorov@mail.ioffe.ru
Keywords: Gene expression, Melting, Nucleic acid annealing, Reductions, Semiconductor

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?