• USA Home
  • 75515 - Orotic acid zinc salt dihydrate

EMAIL THIS PAGE TO A FRIEND
75515 Sigma-Aldrich

Orotic acid zinc salt dihydrate

≥98.0%

  • CAS Number 68399-76-8

  • Empirical Formula (Hill Notation) C10H6N4O8Zn · 2H2O

  • Molecular Weight 411.60

  •  EC Number 269-988-0

  •  MDL number MFCD00167357

  •  PubChem Substance ID 57652569

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C9 to C11, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
Quality Level   100
assay   ≥98.0%
SMILES string   O.O.O=C1NC(=O)C=C(N1)C(=O)O[Zn]OC(=O)C2=CC(=O)NC(=O)N2
InChI   1S/2C5H4N2O4.2H2O.Zn/c2*8-3-1-2(4(9)10)6-5(11)7-3;;;/h2*1H,(H,9,10)(H2,6,7,8,11);2*1H2;/q;;;;+2/p-2
InChI key   KAXHRUFHOVCLAS-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?