• USA Home
  • 756695 - Thallium(III) chloride hydrate

EMAIL THIS PAGE TO A FRIEND
756695 Sigma-Aldrich

Thallium(III) chloride hydrate

98%

  • Empirical Formula (Hill Notation) TlCl3 · xH2O

  •  MDL number MFCD00150365

  •  PubChem Substance ID 329766551

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Thallium Catalysts More...
assay   98%
mp   155 °C (lit.)
SMILES string   O.Cl[Tl](Cl)Cl
InChI   1S/3ClH.H2O.Tl/h3*1H;1H2;/q;;;;+3/p-3
InChI key   SWGLTBCHNXCQPK-UHFFFAOYSA-K

Description

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1707 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?