EMAIL THIS PAGE TO A FRIEND
757586 Aldrich

Sodium nitrate

anhydrous, 99.995% trace metals basis

Synonym: Chile niter, Chile salpeter, Chile saltpeter, Soda niter, Nitric acid, sodium salt

Purchase

Properties

Related Categories Crystal Grade Inorganics, Essential Chemicals, Materials Science, Metal and Ceramic Science, Research Essentials,
grade   anhydrous
InChI Key   VWDWKYIASSYTQR-UHFFFAOYSA-N
assay   99.995% trace metals basis
form   solid
mp   306 °C (dec.) (lit.)
solubility   water: soluble 874 g/L at 20 °C

Description

Packaging

Very hygroscopic. Handle under inert conditions or dry box.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1498 5.1 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Articles

Nanostructured Materials Through Ultrasonic Spray Pyrolysis | Technical Article

Jin Ho Bang,1,2 Yuri T. Didenko,3 Richard J. Helmich,1,4 and Kenneth S. Suslick1* 1School of Chemical Sciences, University of Illinois at Urbana-Champaign, 600 S. Mathews Av., Urbana, IL 61801 USA 2C...
Keywords: Antimicrobials, Cancer, Catalysis, Characterizations, Chemical reactions, Condensations, Deposition, Diffusion, Electronics, Evaporation, Flow cytometry, Nanomaterials, Oxidations, Photovoltaics, Precipitation, Purification, Reductions, Semiconductor, Separation, Solar cells, Solvents, Thin film deposition

Nanostructured Materials Through Ultrasonic Spray Pyrolysis

Advances in materials have often been led by the development of new synthetic methods that provide control over size, morphology and structure. The preparation of materials in a scalable and continuo...
Jin Ho Bang, Yuri T. Didenko, Richard J. Helmich, and Kenneth S. Suslick
Material Matters, Volume 7, Number 2, 15–20
Keywords: Antimicrobials, Cancer, Catalysis, Characterizations, Chemical reactions, Condensations, Deposition, Diffusion, Electronics, Evaporation, Flow cytometry, Nanomaterials, Oxidations, Photovoltaics, Precipitation, Purification, Reductions, Semiconductor, Separation, Solar cells, Solvents, Thin film deposition

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?