• USA Home
  • 759333 - 2-Chloro-3-fluoropyridine-4-carboxaldehyde

EMAIL THIS PAGE TO A FRIEND
759333 Sigma-Aldrich

2-Chloro-3-fluoropyridine-4-carboxaldehyde

97%

Synonym: 2-Chloro-3-fluoro-4-formylpyridine, 2-Chloro-3-fluoroisonicotinaldehyde

Purchase

Properties

Related Categories Building Blocks, C7, C7 to C8, Chemical Synthesis, Halogenated Heterocycles,
InChI Key   RWSUDYQQHLWWDK-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.543
density   1.424 g/mL at 25 °C
storage temp.   −20°C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
226.4 °F
Flash Point(C) 
108 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?