• USA Home
  • 761370 - (Acetonitrile)[2-(dicyclohexylphosphino)3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl] gold(I) hexafluoroantimonate

EMAIL THIS PAGE TO A FRIEND
761370 Sigma-Aldrich

(Acetonitrile)[2-(dicyclohexylphosphino)3,6-dimethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl] gold(I) hexafluoroantimonate

  • CAS Number 1798801-37-2

  • Empirical Formula (Hill Notation) C37H56AuF6NO2PSb

  • Molecular Weight 1010.54

  •  PubChem Substance ID 329766930

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
mp   193-242 °C (decomposition)
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
SMILES string   CC#[N+][Au].F[Sb-](F)(F)(F)(F)F.COc1ccc(OC)c(c1P(C2CCCCC2)C3CCCCC3)-c4c(cc(cc4C(C)C)C(C)C)C(C)C
InChI   1S/C35H53O2P.C2H3N.Au.6FH.Sb/c1-23(2)26-21-29(24(3)4)33(30(22-26)25(5)6)34-31(36-7)19-20-32(37-8)35(34)38(27-15-11-9-12-16-27)28-17-13-10-14-18-28;1-2-3;;;;;;;;/h19-25,27-28H,9-18H2,1-8H3;1H3;;6*1H;/q;;+1;;;;;;;+5/p-6
InChI key   XRZFREZYCQUNTH-UHFFFAOYSA-H

Description

Packaging

100, 500 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?