• USA Home
  • 762547 - 2-Bromo-4-chloropyridine-3-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
762547 Sigma-Aldrich

2-Bromo-4-chloropyridine-3-carboxylic acid

97%

Synonym: 2-Bromo-4-chloronicotinic acid

Purchase

Properties

Related Categories Building Blocks, C7, C7 to C8, Chemical Synthesis, Halogenated Heterocycles,
InChI Key   FICKTDRPOOIPDP-UHFFFAOYSA-N
assay   97%
mp   145-150 °C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?