• USA Home
  • 762954 - 1-Methyl-2-(2-diisopropylphosphinophenyl)-1H-benzoimidazole

EMAIL THIS PAGE TO A FRIEND
762954 Sigma-Aldrich

1-Methyl-2-(2-diisopropylphosphinophenyl)-1H-benzoimidazole

97%

  • CAS Number 1357398-61-8

  • Empirical Formula (Hill Notation) C20H25N2P

  • Molecular Weight 324.40

  •  MDL number MFCD22417225

  •  PubChem Substance ID 329767013

  •  NACRES NA.22

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
assay   97%
mp   147-152 °C
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
SMILES string   CC(C)P(C(C)C)c1ccccc1-c2nc3ccccc3n2C
InChI   1S/C20H25N2P/c1-14(2)23(15(3)4)19-13-9-6-10-16(19)20-21-17-11-7-8-12-18(17)22(20)5/h6-15H,1-5H3
InChI key   RGYYYYMIZGKBRJ-UHFFFAOYSA-N

Description

Packaging

2 g in glass bottle

500 mg in glass bottle

Application

Phosphine ligand for Suzuki coupling of aryl mesylates.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?