• USA Home
  • 763241 - 1-Methyl-2-(2-dicyclohexylphosphinophenyl)-1H-benzoimidazole

EMAIL THIS PAGE TO A FRIEND
763241 Sigma-Aldrich

1-Methyl-2-(2-dicyclohexylphosphinophenyl)-1H-benzoimidazole

95%

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
assay   95%
mp   153-158 °C
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
SMILES string   Cn1c(nc2ccccc12)-c3ccccc3P(C4CCCCC4)C5CCCCC5
InChI   1S/C26H33N2P/c1-28-24-18-10-9-17-23(24)27-26(28)22-16-8-11-19-25(22)29(20-12-4-2-5-13-20)21-14-6-3-7-15-21/h8-11,16-21H,2-7,12-15H2,1H3
InChI key   IBOIHFPCQHFOOP-UHFFFAOYSA-N

Description

Packaging

2 g in glass bottle

500 mg in glass bottle

Application

Phosphine ligand for Suzuki coupling of aryl mesylates.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?