• USA Home
  • 763608 - Manganese nickel carbonate

EMAIL THIS PAGE TO A FRIEND
763608 Aldrich

Manganese nickel carbonate Green Alternative

Ni0.25Mn0.75CO3, 99.99% trace metals basis (excluding Mg)

Synonym: Nickel manganese carbonate

  • CAS Number 911138-32-4

  • Linear Formula Mn0.75Ni0.25CO3

  • Molecular Weight 115.89

Purchase

Properties

Related Categories Alternative Energy, Battery Electrode Materials, Cathode Materials, Chemical Synthesis, Electrode Materials,
assay   99.99% trace metals basis (excluding Mg)
form   powder
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Description

Application

A convenient precursor for the preparation of electrode materials for Li batteries. The material is mixed with Li-salts and decomposed at high temperature to form mixed metal oxides; example Lithium nickel manganese oxide. Lithium nickel manganese oxide is a high-energy and high power electrode material for next generation lithium rechargeable batteries.

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Protocols & Articles

Articles

Electrode Materials for Lithium Ion Batteries

In 2010, the rechargeable lithium ion battery market reached ~$11 billion and continues to grow.1 Current demand for lithium batteries is dominated by the portable electronics and power tool industri...
Kinson C. Kam and Marca M. Doeff
Material Matters, 2012 v7, n4
Keywords: Applications, Atomic absorption spectroscopy, Automotive, Degradations, Electronics, Industries, Ion Exchange, Legal, Oxidations, Precipitation, Purification, Reductions, Renewable energy, Search, Substitutions

Electrolyte Reagents for Lithium-Ion Batteries

Introduction Battery grade electrolytes Solvents and additives Preparation of customized battery grade electrolyte solutions Materials References
Dr. Koji Abe
Advanced Energy Materials R&D Center, Chemicals Company, UBE Industries, Ltd.
Keywords: Acid value, Industries, Solvents

Nanostructured Olivine-based Cathode Materials for Lithium-ion Batteries

Vishwanathan Ramar and Palani Balaya* Department of Mechanical Engineering National University of Singapore, Singapore-117576 *Email: mpepb@nus.edu.sg
Vishwanathan Ramar, Palani Balaya
Material Matters, 2016, 11.1, 23
Keywords: Diffraction, Microscopy, Nanomaterials, Oxidations, Redox Reactions, Reductions, Renewable energy, Scanning electron microscopy, Solvents, Substitutions, Transmission electron microscopy, X-Ray diffraction

Olivine-Type Cathode Materials for Lithium-Ion Batteries

Izumi Taniguchi Department of Chemical Engineering Graduate School of Science and Engineering, Tokyo Institute of Technology 12-1, Ookayama-2, Meguro-ku, Tokyo 152-8552, Japan Email: taniguchi.i.aa@m...
Keywords: Atomic absorption spectroscopy, Deposition, Diffraction, Microscopy, Precipitation, Redox Reactions, Reductions, Scanning electron microscopy, Sol-gel, Spectroscopy, Transmission electron microscopy, X-Ray diffraction

Safer High-performance Electrodes, Solid Electrolytes, and Interface Reactions for Lithium-Ion Batteries

Yves J. Chabal, Kyeongjae Cho, and Christopher L. Hinkle* (Roberto C. Longo, K. C. Santosh, Amandeep K. Sra, David E. Arreaga-Salas, and Katy Roodenko; not pictured) Department of Materials Science a...
Keywords: Absorption, Chemical reactions, Deposition, Diffusion, Electronics, Infrared spectroscopy, Lithiations, Materials Science, Oxidations, Solvents, Spectroscopy, transformation

Scaling Up High-Energy Cathode Materials for Electric Vehicles

Young Ho Shin, Ozgenur Kahvecioglu Feridun, Gregory Krumdick Materials Engineering Research Facility, Energy Systems Division, Argonne National Laboratory 9700 S. Cass Avenue, Argonne, IL, 60439 Emai...
Keywords: Crystallization, Diffraction, Lithiations, Microscopy, Oxidations, Precipitation, Scanning electron microscopy, X-Ray diffraction

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?