• USA Home
  • 763993 - 1′-Hydroxymidazolam-13C3

EMAIL THIS PAGE TO A FRIEND
763993 Aldrich

1′-Hydroxymidazolam-13C3

99 atom % 13C, 98% (CP)

  • Empirical Formula (Hill Notation) 13C3C15H13ClFN3O

  • Molecular Weight 344.74

  •  PubChem Substance ID 329767091

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, G-H, Metabolites, Metabolites and Bioactive Compounds, Stable Isotopes More...
InChI Key   QHSMEGADRFZVNE-BHKULXQMSA-N
isotopic purity   99 atom % 13C
assay   98% (CP)
mass shift   M+3

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis


Stable Isotope
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?