• USA Home
  • 765023 - 2,6-Bis(trimethyltin)-4,8-bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b′]dithiophene

EMAIL THIS PAGE TO A FRIEND
765023 Sigma-Aldrich

2,6-Bis(trimethyltin)-4,8-bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b′]dithiophene

99% (HPLC)

Synonym: BDT-ditin

Purchase

Properties

Related Categories 3D Printing Materials for Research and Development, Materials Science, Organic Semiconductors for 3D Printing, Organic and Printed Electronics, Synthetic Tools and Reagents,
InChI Key   XXMOZDBOAIICDA-UHFFFAOYSA-N
assay   99% (HPLC)
form   powder
mp   70-75 °C
storage temp.   2-8°C

Description

Application

Used as an electron donor in Polymer Solar Cells (PSC).

Packaging

1, 5 g in glass bottle

Features and Benefits

Using BDT-based polymer leads to high power conversion efficiency (PCE) of around 7%-8% in polymer solar cells.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3146 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

3D Printing of Carbon Fiber-Reinforced Composites

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries, most typically using a layer-by-layer fabrication process. 3D pri...
Zhenyu Bo* (Ph.D Candidate at Northwestern University) and Jia Choi*, PhD, Product Manager


*Materials Science Product Management Team, MilliporeSigma, Milwaukee, WI.
Keywords: Deposition, Nanomaterials, Nanotubes

Organic Photovoltaic Applications for IoT, Architecture, and Wearables

1Merck Chemicals Ltd, Chilworth Technical Centre, University Parkway, Southampton. SO16 7QD, U.K. 2Merck KGaA, Frankfurter Str. 250, 64293 Darmstadt I Germany 3Merck Selbstmedikation GmbH, Frankfurte...
Graham Morse,1 Richard Harding,1 Nicolas Blouin,2 Hannah Buerckstrummer2
Not pictured: Agnieszka Pron,1 Stephan Wieder,2 David Mueller,2 and Stephane Berny3
Material Matters, 2017, 12.3
Keywords: Absorption, Alternative energy, Catalysis, Coupling reactions, Cross couplings, Deposition, Environmental, Indicators, Industries, PAGE, Photovoltaics, Polymerization reactions, Positron Emission Tomography, Purification, Safety industry, Semiconductor, Separation, Solar cells, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?