EMAIL THIS PAGE TO A FRIEND
765562 Sigma-Aldrich

Gold nanoparticles

40 nm diameter, carboxylic acid functionalized, PEG 5000 coated, OD 50, dispersion in H2O

Synonym: Au NP COOH, Gold Colloid, Gold nanoparticles COOH functionalized

Purchase

Properties

Related Categories Bioelectronics, Biosensing and Bioimaging, Functionalized Gold Nanoparticles, Gold Nanoparticles, Materials Science,
material   PEG 5000
form   dispersion in H2O
  nanoparticles
packaging   poly bottle of 1 mL
OD   50
diameter   40 nm
pH   6.0-8.0 (25 °C)
solubility   water: miscible
density   1.00 g/cm3
λmax   530 nm
functional group   carboxyl
storage temp.   2-8°C

Description

General description

Gold nanoparticles (AuNPs) ranging from 2-200nm in diameter find applications in diverse fields. They show excellent biocompatibility and attractive physicochemical properties. Poly (ethylene) glycol (PEG) surface coatings on gold nanoparticles help in the reduction of protein adsorption, reduce nonspecific interactions with cells and greatly improve the pharmokinetics of these nanoparticles and reduce aggregation of the nanoparticles. PEG layer density depends inversely on the AuNP curvature. Denser coating was possible on smaller AuNPs. Carboxylic acid functionalized GNPs form conjugates with various protein molecules due to the electrostatic attraction between the negatively charged carboxylate groups and positively charged amino groups of protein. Since it adheres to cell membranes, it may find use in cellular and intracellular targeting in targeted drug delivery applications and also used in biodistribution studies.

Application

Several other specific applications have been listed below:
• Since they adhere to cell membranes, AuNPs may find use in cellular and intracellular targeting in targeted drug delivery applications and may also be used in biodistribution studies.
• AuNPs may also be used in photothermal therapy and radiotherapy.
• AuNPs ranging from 10 to 250 nm, were studied to have been absorbed primarily by liver and spleen, when injected intravenously in rats.Whereas, 10 nm nanoparticles were distributed more broadly into various organs.
•  Carboxylic acid end-group binds with various proteins and can be used in targeted drug delivery and gene therapy.

Functionalized Particles have been extensively used to study cellular uptake of nanoparticles and targeted drug delivery.

Packaging

1 mL in poly bottle

Features and Benefits

The PEG coating decreases the cytotoxicity and increases efficiency of GNPs in targeted drug delivery. PEG increases the stability of the nanoparticles and prevents agglomeration.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Increasing the Sensitivity of Lateral Flow Diagnostic Assays with Ultra-Bright Gold Nanoshell Reporters

Lateral flow assays (also known as immunochromatographic assays) detect or quantify biomolecules in complex samples including blood, urine, saliva, or other fluids to diagnose a variety of medical co...
Keywords: Absorption, Adsorption, Bacterial conjugations, Centrifugation, Deposition, Detergents, Diagnostic, Filtration, Flow cytometry, Hormones, Hydration reaction, Immobilization, Ligands, Microscopy, Raman spectroscopy, Scanning electron microscopy, Spectroscopy, Transmission electron microscopy

Magnetic Nanoparticles for Cancer Theranostics

Saumya Nigam,1 Lina Pradhan,2 and D. Bahadur*3 1IITB-Monash Research Academy, IIT Bombay, Powai, Mumbai, India 2Centre for Research in Nanotechnology & Science IIT Bombay, Powai, Mumbai, India 3Depar...
Keywords: Apoptosis, Bacterial conjugations, Cancer, Diagnostic, Magnetic resonance imaging, Materials Science, Nanomaterials, Nanotechnology, Reductions

Micro- and Nanofabricated Biomaterials for the Study of Cells and Microtissues

Phin Peng Lee, Karen E. Samy, Miquella G. Chavez, Alec Cerchiari, Tejal A. Desai* Department of Bioengineering and Therapeutic Sciences, University of California San Francisco, San Francisco, CA 9415...
Keywords: Absorption, Adhesion, Amplification, Apoptosis, Biomaterials, Cancer, Cell culture, Cell proliferation, Diffusion, Immobilization, Indicators, Infrared spectroscopy, Ligands, Magnetic resonance imaging, Materials Science, Microscopy, Molecular biology, Nanotechnology, Semiconductor, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?