• USA Home
  • 767689 - (2-Biphenyl)di-1-adamantylphosphine

EMAIL THIS PAGE TO A FRIEND
767689 Sigma-Aldrich

(2-Biphenyl)di-1-adamantylphosphine

97%

Synonym: [1,1′-Biphenyl]-2-yldi[(3S,5S,7S)-1-adamantanyl]phosphine

  • CAS Number 224311-55-1

  • Empirical Formula (Hill Notation) C32H39P

  • Molecular Weight 454.63

  •  MDL number MFCD28100470

  •  PubChem Substance ID 329767331

  •  NACRES NA.22

Purchase

Properties

Related Categories Buchwald Ligands and Complexes, Buchwald and Related Ligands, Catalysis and Inorganic Chemistry, Chemical Synthesis, Cross-Coupling,
assay   97%
mp   220-225 °C
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
reaction type: Cross Couplings
SMILES string   [H][C@@]1(C2)C[C@]3([P@@](C4=C(C5=CC=CC=C5)C=CC=C4)[C@]67C[C@]8([H])C[C@](C[C@](C8)([H])C7)([H])C6)C[C@@]2([H])C[C@@](C1)([H])C3
InChI   1S/C32H39P/c1-2-6-28(7-3-1)29-8-4-5-9-30(29)33(31-16-22-10-23(17-31)12-24(11-22)18-31)32-19-25-13-26(20-32)15-27(14-25)21-32/h1-9,22-27H,10-21H2/t22-,23+,24-,25-,26+,27-,31-,32-
InChI key   MSKLBRMTWIUCEQ-ZCTKIPCOSA-N

Description

Packaging

1 g in glass bottle

250 mg in glass bottle

Application

Phosphine ligand for cross coupling.

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?