• USA Home
  • 768197 - (Acetonitrile){4-[2-di(1-adamantyl)phosphino]phenylmorpholine}gold(I) hexafluoroantimonate

EMAIL THIS PAGE TO A FRIEND
768197 Sigma-Aldrich

(Acetonitrile){4-[2-di(1-adamantyl)phosphino]phenylmorpholine}gold(I) hexafluoroantimonate

Synonym: Mor-DalPhos-AuMeCN SbF6

  • Empirical Formula (Hill Notation) C32H45AuF6N2OPSb

  • Molecular Weight 937.40

  •  PubChem Substance ID 329767362

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Gold Catalysts, Gold Complexes
reaction suitability   core: gold
  reagent type: catalyst
mp   150-156 °C (decomposition)
SMILES string   CC#[N+][Au].F[Sb-](F)(F)(F)(F)F.C1CN(CCO1)c2ccccc2P(C34CC5CC(CC(C5)C3)C4)C67CC8CC(CC(C8)C6)C7
InChI   1S/C30H42NOP.C2H3N.Au.6FH.Sb/c1-2-4-28(27(3-1)31-5-7-32-8-6-31)33(29-15-21-9-22(16-29)11-23(10-21)17-29)30-18-24-12-25(19-30)14-26(13-24)20-30;1-2-3;;;;;;;;/h1-4,21-26H,5-20H2;1H3;;6*1H;/q;;+1;;;;;;;+5/p-6
InChI key   WEBSRRMIVVCIAD-UHFFFAOYSA-H

Description

Packaging

100, 500 mg in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Stradiotto Group – Professor Product Portal

From our library of Articles, Sigma-Aldrich presents Stradiotto Group – Professor Product Portal
Keywords: Aminations, Arylations, Buchwald-Hartwig amination, Catalysis, Hydroaminations, Ligands

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?