EMAIL THIS PAGE TO A FRIEND
77005 Sigma-Aldrich

2-Pentenenitrile

technical, ~70% (GC)

Purchase

Properties

grade   technical
assay   ~70% (GC)
refractive index   n20/D 1.422
bp   122-127 °C (lit.)
density   0.821 g/mL at 20 °C (lit.)
SMILES string   CC\C=C\C#N
InChI   1S/C5H7N/c1-2-3-4-5-6/h3-4H,2H2,1H3/b4-3+
InChI key   ISBHMJZRKAFTGE-ONEGZZNKSA-N

Description

Packaging

1 L in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3275 3(6.1) / PGII
WGK Germany 
3
Flash Point(F) 
75.2 °F
Flash Point(C) 
24 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?