• USA Home
  • 77082 - (S)-1,4-Dibromo-2-butanol

EMAIL THIS PAGE TO A FRIEND
77082 Sigma-Aldrich

(S)-1,4-Dibromo-2-butanol

≥95.0% (sum of enantiomers, GC)

  • CAS Number 64028-90-6

  • Empirical Formula (Hill Notation) C4H8Br2O

  • Molecular Weight 231.91

  •  Beilstein/REAXYS Number 5237760

  •  MDL number MFCD00672882

  •  PubChem Substance ID 24887121

Purchase

Properties

assay   ≥95.0% (sum of enantiomers, GC)
mp   33-36 °C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Chiral Alcohols

We offer a wide range of chiral alcohols to meet their ever-growing demand. These useful reagents may serve both as starting materials in the synthesis of single-stereoisomer drugs or intermediates, ...
ChemFiles 2005, 5.4, 11.
Keywords: Diabetes, Nuclear magnetic resonance spectroscopy, Resolving agents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?