• USA Home
  • 771120 - Dimethylaminophenylchlorophosphine

EMAIL THIS PAGE TO A FRIEND
771120 Sigma-Aldrich

Dimethylaminophenylchlorophosphine

95%

Synonym: 1-Chloro-N,N-dimethyl-1-phenylphosphinamine

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
assay   95%
impurities   <5% Bis(dimethylamino)phenylphosphine
density   1.117 g/mL at 25 °C
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
storage temp.   2-8°C
SMILES string   CN(C)P(Cl)c1ccccc1
InChI   1S/C8H11ClNP/c1-10(2)11(9)8-6-4-3-5-7-8/h3-7H,1-2H3
InChI key   XAMVYCVKUOBKPW-UHFFFAOYSA-N

Description

Packaging

2 g in glass bottle

500 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
190.4 °F
Flash Point(C) 
88 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?