• USA Home
  • 77281 - Heptadecafluorooctanesulfonic acid lithium salt solution

EMAIL THIS PAGE TO A FRIEND
77281 Aldrich

Heptadecafluorooctanesulfonic acid lithium salt solution

technical, ~25% in H2O (T)

Synonym: Lithium heptadecafluoro-1-octanesulfonate solution, Perfluorooctanesulfonic acid lithium salt

  • CAS Number 29457-72-5

  • Linear Formula CF3(CF2)7SO3Li

  • Molecular Weight 506.06

  •  Beilstein Registry Number 4346692

  •  MDL number MFCD00467733

  •  PubChem Substance ID 24887170

Purchase

Properties

description   anionic
grade   technical
concentration   ~25% in H2O (T)
refractive index   n20/D 1.334

Description

Other Notes

Anionic fluorocarbon surfactant

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?