• USA Home
  • 773042 - Potassium benzothiophene-3-trifluoroborate

EMAIL THIS PAGE TO A FRIEND
773042 Sigma-Aldrich

Potassium benzothiophene-3-trifluoroborate

95%

Synonym: Potassium 3-benzothiophenetrifluoroborate

Purchase

Properties

Related Categories Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Trifluoroborate Salts, Organometallic Reagents, Trifluoroborate Salts More...
assay   95%
mp   178.5-186.8 °C
SMILES string   F[B-](F)(F)C1=CSC2=C1C=CC=C2.[K+]
InChI   1S/C8H5BF3S.K/c10-9(11,12)7-5-13-8-4-2-1-3-6(7)8;/h1-5H;/q-1;+1
InChI key   CMEOIXLPVLKJDC-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?