EMAIL THIS PAGE TO A FRIEND
773964 Sigma-Aldrich

Aluminum

ingot, L × W 10 mm × 7 mm, 99.99995% trace metals basis

Purchase

Properties

Related Categories Al, Aluminum, Materials Science, Metal and Ceramic Science, Metals,
InChI Key   XAGFODPZIPBFFR-UHFFFAOYSA-N
assay   99.99995% trace metals basis
form   ingot
autoignition temp.   1400 °F
resistivity   2.6548 μΩ-cm
L × W   10 mm × 7 mm
weight   1.5 g
bp   2460 °C (lit.)
mp   660.37 °C (lit.)
density   2.7 g/mL at 25 °C (lit.)

Description

Application

Ultra high purity aluminum is an interesting material for use in many high technology semiconductor applications based on molecular beam epitaxy including IR absorbers and quantum cascade lasers.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles

Articles

Biomedical Implant Devices Fabricated from Low Young’s Modulus Titanium Alloys Demonstrating High Mechanical Biocompatibility

Mitsuo Niinomi, Masasaki Nakai* Institute for Materials Research, Tohoku University 2-1-1, Katahira, Aoba-ku, Sendai 8577, Japan *not pictured Email: niinomi@imr.tohoku.ac.jp

Responsive Rare-Earth Materials and their Applications

Yaroslav Mudryk Ames Laboratory of U.S. Department of Energy, Iowa State University Ames, Iowa 50011 USA Email: slavkomk@ameslab.gov
Yaroslav Mudryk
Material Matters, 2016, 11.4
Keywords: Diffraction, PAGE, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?