EMAIL THIS PAGE TO A FRIEND
774294 Aldrich

Propylene sulfate Green Alternative

≥99%

Synonym: 1,2-Propanediol cyclic sulfate, 4-Methyl-1,3,2-dioxathiolane-2,2-dioxide, 4-Methyl-2,2-dioxo-1,3,2-dioxathiolane

Purchase

Properties

Related Categories Alternative Energy, Battery Electrolyte Materials, Chemical Synthesis, Electrolytes, Enabling Products,
InChI Key   OQXNUCOGMMHHNA-UHFFFAOYSA-N
assay   ≥99%
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
refractive index   n/D 1.419
bp   187-198 °C/760 mmHg (decomposition)
density   1.408 g/mL
Featured Industry   Battery Manufacturing

Description

Application

Propylene sulfate is used as an additive in lithium ion batteries to improve high temperature performance and cyclic performance. It also suppresses initial irreversible capacity loss in graphite electrodes.

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles

Articles

Electrolyte Reagents for Lithium-Ion Batteries

Introduction Battery grade electrolytes Solvents and additives Preparation of customized battery grade electrolyte solutions Materials References
Dr. Koji Abe
Advanced Energy Materials R&D Center, Chemicals Company, UBE Industries, Ltd.
Keywords: Acid value, Industries, Solvents

Nanostructured Olivine-based Cathode Materials for Lithium-ion Batteries

Vishwanathan Ramar and Palani Balaya* Department of Mechanical Engineering National University of Singapore, Singapore-117576 *Email: mpepb@nus.edu.sg
Vishwanathan Ramar, Palani Balaya
Material Matters, 2016, 11.1, 23
Keywords: Diffraction, Microscopy, Nanomaterials, Oxidations, Redox Reactions, Reductions, Renewable energy, Scanning electron microscopy, Solvents, Substitutions, Transmission electron microscopy, X-Ray diffraction

Olivine-Type Cathode Materials for Lithium-Ion Batteries

Izumi Taniguchi Department of Chemical Engineering Graduate School of Science and Engineering, Tokyo Institute of Technology 12-1, Ookayama-2, Meguro-ku, Tokyo 152-8552, Japan Email: taniguchi.i.aa@m...
Keywords: Atomic absorption spectroscopy, Deposition, Diffraction, Microscopy, Precipitation, Redox Reactions, Reductions, Scanning electron microscopy, Sol-gel, Spectroscopy, Transmission electron microscopy, X-Ray diffraction

Safer High-performance Electrodes, Solid Electrolytes, and Interface Reactions for Lithium-Ion Batteries

Yves J. Chabal, Kyeongjae Cho, and Christopher L. Hinkle* (Roberto C. Longo, K. C. Santosh, Amandeep K. Sra, David E. Arreaga-Salas, and Katy Roodenko; not pictured) Department of Materials Science a...
Keywords: Absorption, Chemical reactions, Deposition, Diffusion, Electronics, Infrared spectroscopy, Lithiations, Materials Science, Oxidations, Solvents, Spectroscopy, transformation

Scaling Up High-Energy Cathode Materials for Electric Vehicles

Young Ho Shin, Ozgenur Kahvecioglu Feridun, Gregory Krumdick Materials Engineering Research Facility, Energy Systems Division, Argonne National Laboratory 9700 S. Cass Avenue, Argonne, IL, 60439 Emai...
Keywords: Crystallization, Diffraction, Lithiations, Microscopy, Oxidations, Precipitation, Scanning electron microscopy, X-Ray diffraction

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?