• USA Home
  • 776394 - Lanthanum yttrium germanate, LaYGe

EMAIL THIS PAGE TO A FRIEND
776394 Sigma-Aldrich

Lanthanum yttrium germanate, LaYGe

Green Alternative

99.995%

Synonym: Germanium lanthanum yttrium oxide, LaYGe

Purchase

Properties

Related Categories Alternative Energy, Chemical Synthesis, Electrolyte Materials, Electrolytes, Enabling Products,
assay   99.995%
form   powder
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Description

Application

Lanthanum Yttrium Germanate is an apatite-type rare earth germanate which has gathered considerable attention recently for its high oxide ion conductivity and finds applications as electrolyte material for Solid Oxide Fuel Cells.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?