• USA Home
  • 777110 - Lithium cobalt phosphate

EMAIL THIS PAGE TO A FRIEND
777110 Aldrich

Lithium cobalt phosphate Green Alternative

99.9% trace metals basis, battery grade

Synonym: Cobalt lithium phosphate, LCP

Purchase

Properties

Related Categories Alternative Energy, Battery Electrode Materials, Cathode Materials, Chemical Synthesis, Electrode Materials,
grade   battery grade
InChI Key   SBWRUMICILYTAT-UHFFFAOYSA-K
assay   99.9% trace metals basis
form   powder
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

Description

Application

Lithium cobalt phosphate (LCP) with the phospho-olivine structure is a high voltage cathode material for rechargeable lithium ion batteries. The material retains its structure and crystallinity after repeated charge-discharge cycles. The partically delithiated material shows high thermal stability as well.

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Electrolyte Reagents for Lithium-Ion Batteries

Introduction Battery grade electrolytes Solvents and additives Preparation of customized battery grade electrolyte solutions Materials References
Dr. Koji Abe
Advanced Energy Materials R&D Center, Chemicals Company, UBE Industries, Ltd.
Keywords: Acid value, Industries, Solvents

Nanostructured Olivine-based Cathode Materials for Lithium-ion Batteries

Vishwanathan Ramar and Palani Balaya* Department of Mechanical Engineering National University of Singapore, Singapore-117576 *Email: mpepb@nus.edu.sg
Vishwanathan Ramar, Palani Balaya
Material Matters, 2016, 11.1, 23
Keywords: Diffraction, Microscopy, Nanomaterials, Oxidations, Redox Reactions, Reductions, Renewable energy, Scanning electron microscopy, Solvents, Substitutions, Transmission electron microscopy, X-Ray diffraction

Olivine-Type Cathode Materials for Lithium-Ion Batteries

Izumi Taniguchi Department of Chemical Engineering Graduate School of Science and Engineering, Tokyo Institute of Technology 12-1, Ookayama-2, Meguro-ku, Tokyo 152-8552, Japan Email: taniguchi.i.aa@m...
Keywords: Atomic absorption spectroscopy, Deposition, Diffraction, Microscopy, Precipitation, Redox Reactions, Reductions, Scanning electron microscopy, Sol-gel, Spectroscopy, Transmission electron microscopy, X-Ray diffraction

Safer High-performance Electrodes, Solid Electrolytes, and Interface Reactions for Lithium-Ion Batteries

Yves J. Chabal, Kyeongjae Cho, and Christopher L. Hinkle* (Roberto C. Longo, K. C. Santosh, Amandeep K. Sra, David E. Arreaga-Salas, and Katy Roodenko; not pictured) Department of Materials Science a...
Keywords: Absorption, Chemical reactions, Deposition, Diffusion, Electronics, Infrared spectroscopy, Lithiations, Materials Science, Oxidations, Solvents, Spectroscopy, transformation

Scaling Up High-Energy Cathode Materials for Electric Vehicles

Young Ho Shin, Ozgenur Kahvecioglu Feridun, Gregory Krumdick Materials Engineering Research Facility, Energy Systems Division, Argonne National Laboratory 9700 S. Cass Avenue, Argonne, IL, 60439 Emai...
Keywords: Crystallization, Diffraction, Lithiations, Microscopy, Oxidations, Precipitation, Scanning electron microscopy, X-Ray diffraction

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?