• USA Home
  • 778419 - 4,5-Dichloro-3H-1,2-dithiol-3-one

EMAIL THIS PAGE TO A FRIEND
778419 Sigma-Aldrich

4,5-Dichloro-3H-1,2-dithiol-3-one

98%

  • CAS Number 1192-52-5

  • Empirical Formula (Hill Notation) C3Cl2OS2

  • Molecular Weight 187.07

  •  Beilstein/REAXYS Number 1342220

  •  EC Number 214-754-5

  •  MDL number MFCD00043025

  •  PubChem Substance ID 329767965

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Other Heterocyclic Building Blocks, S-Containing More...
InChI Key   QGSRKGWCQSATCL-UHFFFAOYSA-N
assay   ≥98% (GC)
  98%
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
392 °F
Flash Point(C) 
200 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?