• USA Home
  • 778613 - 3-Azidotoluene solution

EMAIL THIS PAGE TO A FRIEND
778613 Sigma-Aldrich

3-Azidotoluene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym: 1-Azido-3-methylbenzene solution, 3-Methylphenyl azide solution, m-Tolyl azide solution

  • CAS Number 4113-72-8

  • Empirical Formula (Hill Notation) C7H7N3

  • Molecular Weight 133.15

  •  Beilstein/REAXYS Number 1817552

  •  MDL number MFCD00211419

  •  PubChem Substance ID 329767982

Purchase

Properties

Related Categories Aromatic Azides, Azides, Building Blocks, Chemical Synthesis, Nitrogen Compounds - Organic Building Blocks,
Quality Level   100
assay   ≥95.0% (HPLC)
concentration   ~0.5 M in tert-butyl methyl ether
impurities   ≤2.0% water
storage temp.   −20°C
SMILES string   Cc1cccc(c1)N=[N+]=[N-]
InChI   1S/C7H7N3/c1-6-3-2-4-7(5-6)9-10-8/h2-5H,1H3
InChI key   WRMGCIOUNQGMSR-UHFFFAOYSA-N

Description

Packaging

10 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2398 3 / PGII
WGK Germany 
3
Flash Point(F) 
-27.4 °F
Flash Point(C) 
-33 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?