• USA Home
  • 778745 - (SP)-1-[(R)-tert-Butylphosphinoyl]-2-[(R)-1-(diphenylphosphino)ethyl]ferrocene

EMAIL THIS PAGE TO A FRIEND
778745 Sigma-Aldrich

(SP)-1-[(R)-tert-Butylphosphinoyl]-2-[(R)-1-(diphenylphosphino)ethyl]ferrocene

97%

Synonym: (1R)-1-[(R)-(1,1-Dimethylethyl)phosphinyl]-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene (acc to CAS), JoSPOphos SL-J681-1

  • CAS Number 1221745-90-9

  • Empirical Formula (Hill Notation) C28H32FeOP2

  • Molecular Weight 502.35

  •  PubChem Substance ID 329767991

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, JoSPOphos, Privileged Ligands and Complexes,
assay   ≥97%
  97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2[P@@H](=O)C(C)(C)C)P(c3ccccc3)c4ccccc4
InChI   1S/C23H27OP2.C5H5.Fe/c1-18(21-16-11-17-22(21)26(24)23(2,3)4)25(19-12-7-5-8-13-19)20-14-9-6-10-15-20;1-2-4-5-3-1;/h5-18,26H,1-4H3;1-5H;/t18-;;/m1../s1
InChI key   LOIKSPAEIVMSHK-JPKZNVRTSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?