• USA Home
  • 78151 - 1-Ethyl-2,3-dimethylimidazolium chloride

EMAIL THIS PAGE TO A FRIEND
78151 Sigma-Aldrich

1-Ethyl-2,3-dimethylimidazolium chloride

≥97.0% (T)

Purchase

Properties

Related Categories Chemical Synthesis, Imidazolium ionic liquids, Ionic Liquids, Specialty Synthesis
assay   ≥97.0% (T)
SMILES string   [Cl-].CCn1cc[n+](C)c1C
InChI   1S/C7H13N2.ClH/c1-4-9-6-5-8(3)7(9)2;/h5-6H,4H2,1-3H3;1H/q+1;/p-1
InChI key   NUJYCYLTUXYHQU-UHFFFAOYSA-M
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?