EMAIL THIS PAGE TO A FRIEND
790923 Sigma-Aldrich

Aluminum oxide

nanofiber, diam. × L <20 nm × 500 μm

Synonym: Alumina Nanofibers, Alumina nanowires, Alumina, Aluminum oxide nanowires

Purchase

Properties

Related Categories 13: Al, Materials Science, Nanomaterials, Nanoparticles: Materials by Application, Nanoparticles: Oxides, Nitrides, and Other Ceramics,
InChI Key   TWNQGVIAIRXVLR-UHFFFAOYSA-N
form   nanofiber
diam. × L   <20 nm × 500 μm
surface area   120-150 m2/g (Surface dimension,BET)
mp   2040 °C (lit.)

Description

Application

Aluminum oxide nanofibers have significant potential as support materials in heterogeneous catalysis and have been tested in numerous catalytic systems including methane reforming and Fischer-Tropsch reactions. Alumina nanowires can also serve to reinforce polymer composites and have been shown to directly effect the electronic properties of conductive polymers.,

Packaging

25 g in poly bottle

Legal Information

Product of ANF Technologies

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Molybdenum Disulfide: Understanding Hydrogen Evolution Catalysis

Linyou Cao Department of Materials Science and Engineering Department of Physics North Carolina State University, Raleigh, NC, USA Email: lcao2@ncsu.edu
Keywords: Adsorption, Bacterial conjugations, Catalysis, Chemical industry, Chemical vapor deposition, Degradations, Deposition, Electrochemical reactions, Environmental, Indicators, Industries, Infrared spectroscopy, Materials Science, Nanotubes, Oxidations, Recombination, Renewable energy

Thiolated gold nanoclusters: A new class of photosensitizers

Kevin Stamplecoskie, Yong-Siou Chen, Prashant V. Kamat* Radiation Laboratory and Department of Chemistry and Biochemistry University of Notre Dame, Notre Dame, Indiana, USA *Email: pkamat@nd.edu
Keywords: Absorption, Biochemistry, Diffraction, Ligands, Nanomaterials, Oxidations, Photovoltaics, Reductions, Solar cells, Spectroscopy, X-Ray diffraction

Titania Nanotubes: Synthesis and Applications

Eugen Panaitescu1 and Latika Menon2 Department of Physics Northeastern University, Boston, MA 02115 USA Email: 1e.panaitescu@neu.edu, 2l.menon@neu.edu
Keywords: Adsorption, Agriculture, Antimicrobials, Apoptosis, Automotive, Calorimetry, Cosmetics, Degradations, Deposition, Diffusion, Electrochemical analysis, Environmental, Filtration, Melting, Nanotubes, Oxidations, Photovoltaics, Precipitation, Recombination, Redox Reactions, Reductions, Semiconductor, Separation, Sol-gel, Solar cells, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?