• USA Home
  • 790974 - Lithium iron(II) silicate

EMAIL THIS PAGE TO A FRIEND
790974 Aldrich

Lithium iron(II) silicate Green Alternative

powder, <2 μm particle size, 99.7% trace metals basis

Synonym: Iron lithium silicate, LFS

  • CAS Number 30734-07-7

  • Empirical Formula (Hill Notation) Li2FeSiO4

  • Molecular Weight 161.81

Purchase

Properties

Related Categories Alternative Energy, Battery Electrode Materials, Cathode Materials, Chemical Synthesis, Electrode Materials,
assay   99.7% trace metals basis
form   powder
greener alternative product characteristics   Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
particle size   <2 μm

Description

Application

Li2FeSiO4 (LFS) based materials have attracted a great deal of interest as cathodes for lithium ion batteries because of its excellent thermal stability and good cycling ability. LFS is attracting much attention recently because it is composed of abundant elements and is capable of exchanging >1 Li per redox-active transition metal ion.

General description

Sigma Life Science is committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Electrolyte Reagents for Lithium-Ion Batteries

Introduction Battery grade electrolytes Solvents and additives Preparation of customized battery grade electrolyte solutions Materials References
Dr. Koji Abe
Advanced Energy Materials R&D Center, Chemicals Company, UBE Industries, Ltd.
Keywords: Acid value, Industries, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?