• USA Home
  • 79178 - (Phenylthio)acetonitrile

EMAIL THIS PAGE TO A FRIEND
79178 Sigma-Aldrich

(Phenylthio)acetonitrile

technical, ≥95% (GC)

Synonym: (Phenylmercapto)acetonitrile

Purchase

Properties

grade   technical
assay   ≥95% (GC)
refractive index   n20/D 1.583 (lit.)
  n20/D 1.583
bp   146-147 °C/14 mmHg (lit.)
density   1.142 g/mL at 25 °C (lit.)
SMILES string   N#CCSc1ccccc1

Description

Other Notes

Versatile CH-acidic compound; Michael additions; Vicarious substitution of hydrogen in aryls

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3334
WGK Germany 
WGK 3
RTECS 
AM1900000
Flash Point(F) 
235.4 °F - closed cup
Flash Point(C) 
113 °C - closed cup
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?