• USA Home
  • 79199 - (R)-5-Hydroxy-2-hexanone

EMAIL THIS PAGE TO A FRIEND
79199 Aldrich

(R)-5-Hydroxy-2-hexanone

technical, ≥90% (sum of enantiomers, GC)

  • CAS Number 65709-73-1

  • Empirical Formula (Hill Notation) C6H12O2

  • Molecular Weight 116.16

  •  Beilstein/REAXYS Number 6287241

  •  MDL number MFCD04040054

  •  PubChem Substance ID 24887419

Purchase

Properties

grade   technical
assay   ≥90% (sum of enantiomers, GC)
optical purity   enantiomeric ratio: ≥99:1 (GC)

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?