• USA Home
  • 795275 - Poly(2-methyl-2-oxazoline), hydroxy terminated

EMAIL THIS PAGE TO A FRIEND
795275 Aldrich

Poly(2-methyl-2-oxazoline), hydroxy terminated

average Mn 2,000, PDI <1.2

Synonym: PMOx

Purchase

Properties

Related Categories Hydrophilic Polymers, Materials Science, Poly(2-oxazoline), Poly(2-oxazoline) and Polyethylenimine (PEI), Polymer Science,
InChI Key   OBSKGKGKPGOOTB-UHFFFAOYSA-N
mol wt   average Mn 2,000
PDI   <1.2
storage temp.   2-8°C

Description

Packaging

5 g in glass bottle

Application

Poly(2-methyl-2-oxazoline) is a hydrophilic polymer that is commonly used in biomedical applications. This is a biocompatible polymer with one chain-end functionalized with an OH group. They are commonly used as an amorphous alternative to PEGs.

Price and Availability


The Future of Bioimaging

Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Poly(2-Oxazoline)S: The Versatile Polymer Platform for Biomedicine

Victor R. de la Rosa,1 An Van Den Bulcke,2 and Richard Hoogenboom1* 1Supramolecular Chemistry Group, Department of Organic and Macromolecular Chemistry Ghent University, Krijgslaan 281-S4, 9000 Ghent...
Victor R. de la Rosa, An Van Den Bulcke, and Richard Hoogenboom
Material Matters, 2016, 11.3
Keywords: Adhesion, Adsorption, Amidations, Bacterial conjugations, Biomaterials, Cancer, Catalysis, Click chemistry, Clinical, Cosmetics, Cycloadditions, Degradations, Electron spin resonance, Pharmaceutical, Polymerization reactions, Ring-opening polymerization

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?