EMAIL THIS PAGE TO A FRIEND
796387 Aldrich

Silver nanospheres

20 nm avg. part. size, 0.02 mg/mL in water, BPEI functionalized

  • Empirical Formula (Hill Notation) Ag

  • Molecular Weight 107.87

Purchase

Properties

Related Categories 47: Ag, Biomedical Materials, Materials Science, Nanomaterials, Nanoparticles: Materials by Application,
concentration   0.02 mg/mL in water
avg. part. size   20 nm
density   1.0007 g/mL at 25 °C
absorption   λmax 400 nm
functional group   BPEI
storage temp.   2-8°C

Description

Packaging

25 mL in glass bottle

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications. A resurgence of interest in the utility of silver as a broad based antimicrobial agent has led to the development of hundreds of products that incorporate silver nanoparticles to prevent bacterial growth on surfaces. Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle’s size and shape. The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules, highly efficient thermal absorbers, and nanoscale "antennas" that can amplify the strength of the local electromagnetic field.

General description

Store products away from light at 4-6°C. Short periods at room temperature are acceptable however, lower temperature storage prolongs the shelf life of the product. DO NOT FREEZE.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Webinar - Hydrogels