• USA Home
  • 802808 - 2,6-Difluorobenzylphosphonic acid

EMAIL THIS PAGE TO A FRIEND
802808 Sigma-Aldrich

2,6-Difluorobenzylphosphonic acid

Synonym: 2,6-F2BPA

  • CAS Number 273211-31-7

  • Empirical Formula (Hill Notation) C7H7F2O3P

  • Molecular Weight 208.10

  •  PubChem Substance ID 329768756

Purchase

Properties

Related Categories Materials Science, Micro/NanoElectronics, Phosphonic Acids, Self Assembly and Lithography, Self-Assembly Materials More...
InChI Key   BNNGITGCJOYNHF-UHFFFAOYSA-N

Description

Packaging

250 mg in poly bottle

Application

Tuning of surface energy, wetting and work function of metal oxides.

Safety & Documentation

Safety Information

RIDADR 
UN 3261 8 / PGII
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?