EMAIL THIS PAGE TO A FRIEND
803634 Sigma-Aldrich

Silica nanospheres

160 nm avg. part. size (TEM), 10 % (w/v) in ethanol

  • Empirical Formula (Hill Notation) SiO2

  • Molecular Weight 60.08

Purchase

Description

Application

Traditionally the silica (SiO2) nanoparticles find applications as additives to drugs, cosmetics, printer toners, varnishes, and food etc. In recent years, their use has been extended to biomedical and biotechnological fields as biosensors, biomarkers, DNA and drug delivery agents etc.

Packaging

1 mL in glass bottle

Safety & Documentation

Safety Information

RIDADR 
UN1170 - class 3 - PG 2 - Ethanol, solution

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?