• USA Home
  • 804509 - Potassium 2-isonicotinoyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
804509 Sigma-Aldrich

Potassium 2-isonicotinoyltrifluoroborate

  • CAS Number 1622923-53-8

  • Empirical Formula (Hill Notation) C6H4BF3KNO

  • Molecular Weight 213.01

  •  PubChem Substance ID 329768876

Purchase

Properties

Related Categories Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Trifluoroborate Salts, Organometallic Reagents, Trifluoroborate Salts More...
Quality Level   100
form   powder
mp   239-244 °C
SMILES string   O=C(B(F)(F)F)C1=CC=CC=N1.[K]
InChI   1S/C6H4BF3NO.K.H/c8-7(9,10)6(12)5-3-1-2-4-11-5;;/h1-4H;;
InChI key   SONNVKVCVWXMGV-UHFFFAOYSA-N

Description

General description

Potassium 2-isonicotinoyltrifluoroborate belongs to the class of compounds known as potassium acyltrifluroborates (KATs). This trifluoroborate salt is stable and does not undergo trimerization.

Application

Potassium acyltrifluroborates (KATs) are bench, air and moisture stable reagents for rapid, chemoselective amide bond formations with hydroxylamines. These amide bond forming reactions proceed under aqueous conditions, without the need for coupling reagents or protecting groups. This product was introduced in collaboration with the Bode Research Group.

Packaging

1 g in glass bottle

Other Notes

Rapid Ligations with Equimolar Reactants in Water with the Potassium Acyltrifluoroborate (KAT) Amide Formation
Amide-Forming Ligation of Acyltrifluoroborates and Hydroxylamines in Water
Synthesis of Chemically and Configurationally Stable Monofluoro Acylboronates: Effect of Ligand Structure on their Formation, Properties, and Reactivities

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Jeffrey Bode Group – Professor Product Portal

From our library of Articles, Sigma-Aldrich presents Jeffrey Bode Group – Professor Product Portal
Keywords: Annulations, Asymmetric synthesis, Catalysis, Cross couplings, transformation

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?