EMAIL THIS PAGE TO A FRIEND
808776 Sigma-Aldrich

Gold nanoparticles

40 nm diameter, streptavidin conjugated, buffered in PBS

Synonym: Streptavidin gold

Purchase

Properties

Related Categories 79: Au, Functionalized Gold Nanoparticles, Gold Nanoparticles, Materials Science, Nanomaterials,
mol wt   mol wt 10 kDa (PEG Linker Length)
composition   BSA, 1%
  glycerol, 20 % (v/v)
  PBS, 0.01 M (pH 7.4)
  , 2.14E+11 particles/mL
OD   2.9-3.1
diameter   40 nm
absorption   absorption 530 nm (Peak SPR)
storage temp.   2-8°C

Description

Application

Streptavidin/Gold conjugates are suitable for the use in immunoblotting, light microscopy, and electron microscope applications.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis


Webinar - Hydrogels
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?