• USA Home
  • 81130 - Poly(acrylic acid sodium salt)

EMAIL THIS PAGE TO A FRIEND
81130 Aldrich

Poly(acrylic acid sodium salt)

2,100

Synonym: Poly(sodium acrylate), Sodium polyacrylate

Purchase

Price and Availability


Webinar: Recent Developments in RAFT Agents

The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
WD6826000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?